missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01252730

 View more versions of this product

Catalog No. AA1252730



Description

Description

Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc 2,4-pentanedionate monohydrate
136°C to 138°C (decomposition)
C10H16O5Zn
250 g
Soluble in alcohol; Slightly soluble in water
O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
281.61
281.60 (263.59 Anhydrous)
14363-15-6
1.6 g/mL
MFCD00000035
zinc 2,4-pentanedionate monohydrate
BNHYOPPPENBGNL-SUKNRPLKSA-L
zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
122130627
Crystalline

Chemical Identifiers

14363-15-6
281.61
BNHYOPPPENBGNL-SUKNRPLKSA-L
122130627
O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]
C10H16O5Zn
MFCD00000035
zinc 2,4-pentanedionate monohydrate
zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-860-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only