Please login to your online account to display your discounted pricing

Zinc, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™


Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Quantity 100mL
Packaging Poly Bottle

Manufacturer: avantor performance  579104

 View more versions of this product

Catalog No. 02004583

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 65.39
Name Note Plasma Standard
Molecular Formula Zn
Chemical Name or Material Zinc, 1000μg/mL (0.10% w/v)
Quantity 100mL
Specific Gravity 1L = 1.01kg
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Poly Bottle

Specifications

Chemical Name or Material Zinc, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Zn
Formula Weight 65.39