Please login to your online account to display your discounted pricing

Zinc, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$111.15 - $153.50

Chemical Identifiers

Chemical Name or Material Zinc, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Zn
Formula Weight 65.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004583

 
avantor performance
579104
Poly Bottle 100mL Each for $153.50

02004584

 
avantor performance
647404
Narrow Mouth Poly Bottle 150mL Each for $111.15
Description & Specifications

Specifications

Formula Weight 65.39
Molecular Formula Zn
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Specific Gravity 1L = 1.01kg
Quantity 100mL
Packaging Poly Bottle
Chemical Name or Material Zinc, 1000μg/mL (0.10% w/v)