Please login to your online account to display your discounted pricing

Zimelidine dihydrochloride, Tocris

Selective 5-HT uptake inhibitor

$97.76 - $410.62

Chemical Identifiers

Chemical Name or Material Zimelidine dihydrochloride
Assay Percent Range >98%
Molecular Formula C16H17BrN2·2HCl
Formula Weight 390.15
Synonym (2Z)-3-(4-Bromophenyl)-N, N-dimethyl-3-(3-pyrindinyl)-2-propen-1-amine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

176750

 
r & d systems
RDS176750
50mg Each for $410.62

176710

 
r & d systems
RDS176710
10mg Each for $97.76
Description & Specifications

Specifications

Formula Weight 390.15
Quantity 50mg
Chemical Name or Material Zimelidine dihydrochloride
Assay Percent Range >98%
Molecular Formula C16H17BrN2·2HCl
CAS 60525-15-7
Synonym (2Z)-3-(4-Bromophenyl)-N, N-dimethyl-3-(3-pyrindinyl)-2-propen-1-amine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature