Please login to your online account to display your discounted pricing
Zetex™ and ZetexPlus™ Specialized Gloves and Mittens
$130.42 - $145.70
Specifications
Length (English) 14 in.
Length (Metric) 35.56cm
Size One size
View More Specs
Products
Catalog Number Mfr. No. Temperature (English) Max. Temperature (Metric) Max Color Material Price Quantity    
11-392-54
newtex industries inc
2100009
600°F 316°C White Leather Pair for $145.70
Case of 48 Pair for $6,205.92
11-392-58
newtex industries inc
2100040
2000°F 1093°C Tan Fiberglass Pair for $130.42
Case of 12 Pair for $1,428.96
Description & Specifications
Specifications
Length (English) 14 in.
Length (Metric) 35.56cm
Size One size
Offer the same protection as standard Zetex™ or ZetexPlus™ gloves and mittens but with added features. Zetex ™ gloves with leather palm deliver extra cut resistance. Compared with single-palm ZetexPlus™ mittens, double-palm ZetexPlus™ mittens extend handling time as much as twofold. Aluminized Zetex™ gloves offer dust-free radiant heat reflection.
All styles sold by the pair.