Binder™ Zero Voltage Alarm Output
Binder™ Zero Voltage Alarm Output for temperature and humidity values.
$2,012.46 - $3,440.85
Specifications
For Use With (Application) Temperature and humidity
Type Alarm Output
View More Specs
Products 14
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
08111093 View Documents Binder™
80121001
KT 115 Each for $2,012.46
08111094 View Documents Binder™
80121002
KT 170 Each for $2,012.46
08111092 View Documents Binder™
80120982
KT 53 Each for $2,012.46
08111095 View Documents Binder™
80121082
KB 53, KB 115 Each for $2,012.46
08111096 View Documents Binder™
80121083
KB 720, KB 240, KB 400 Each for $2,012.46
08111098 View Documents Binder™
80121037
KBF 240 Each for $3,440.85
08111099 View Documents Binder™
80121038
KBF 720 Each for $3,440.85
08111100 View Documents Binder™
80121810
KBF 1020 Each for $3,440.85
08111101 View Documents Binder™
80121758
KBF P 240 Each for $3,440.85
08111102 View Documents Binder™
80121762
KBF P 720 Each for $3,440.85
08111103 View Documents Binder™
80121759
KBF LQC 240 Each for $3,440.85
08111104 View Documents Binder™
80121763
KBF LQC 720 Each for $3,440.85
08111106 View Documents Binder™
80121677
KMF 240 Each for $3,440.85
08111107 View Documents Binder™
80121678
KMF 720 Each for $3,440.85
Description
Description
  • Temperature ±2°C, humidity ±5% RH
  • Accessible via 6-pin DIN socket (max. 24 V – 2.5 A)
  • With audible signal that can be switched off
Specifications
Specifications
Temperature and humidity
Alarm Output