missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BINDER™ Zero Voltage Alarm Output

For Use With: KB 53, KB 115

Supplier:  BINDER™ 80121082

Catalog No. 08111095


Description

Description

  • Temperature ±2°C, humidity ±5% RH
  • Accessible via 6-pin DIN socket (max. 24 V – 2.5 A)
  • With audible signal that can be switched off
Specifications

Specifications

Alarm output, zero-voltage - for temperature (±2°C), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), with audible signal that can be switched off
BINDER Chambers KB 53, KB 115
Alarm Output
Temperature and Humidity
SDS
Documents

Documents

Promotions

Promotions