Alfa Aesar™ Zeolite ZSM-5, ammonium
Manufacturer:  Alfa Aesar™ 4588036
 View more versions of this product
Catalog No. AA4588036
Description
Description
  • Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
Specifications
Specifications
Zeolite ZSM-5, ammonium
Al2O5Si
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax®, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
HNPSIPDUKPIQMN-UHFFFAOYSA-N
dioxosilane;oxo(oxoalumanyloxy)alumane
9942228
400m2/g
500g
1318-02-1
MFCD00132601
SiO2:Al2O3, 30:1 mole ratio
O=[Al]O[Al]=O.O=[Si]=O
162.043
Powder
White
Chemical Identifiers
1318-02-1
162.043
HNPSIPDUKPIQMN-UHFFFAOYSA-N
9942228
O=[Al]O[Al]=O.O=[Si]=O
Al2O5Si
MFCD00132601
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax®, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
dioxosilane;oxo(oxoalumanyloxy)alumane
SDS Documents