missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zeolite, type: ZSM-5, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC279570050
Description
AdsorbentSpecifications
Zeolite | |
C14H16N2 | |
1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, 4-2-ethyl-2-indanyl imidazole, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, antisedan, atipamezol spanish, atipamezole, atipamezolum latin, dsstox_cid_29061, unii-03n9u5jaf6 | |
HSWPZIDYAHLZDD-UHFFFAOYSA-N | |
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole | |
71310 | |
400 to 570 Molar ratio SiO2/Al2O3 | |
700ppm max. | |
300 m2/g (BET) min. | |
5kg |
308081-08-5 | |
MFCD00132601 | |
Solubility in water: insoluble | |
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3 | |
212.296 | |
Crystalline Powder | |
10 % max. (1000°C) | |
Solubility in water: insoluble. | |
Yellow |
Chemical Identifiers
308081-08-5 | |
212.296 | |
HSWPZIDYAHLZDD-UHFFFAOYSA-N | |
71310 | |
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3 |
C14H16N2 | |
MFCD00132601 | |
1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, 4-2-ethyl-2-indanyl imidazole, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, antisedan, atipamezol spanish, atipamezole, atipamezolum latin, dsstox_cid_29061, unii-03n9u5jaf6 | |
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves
GHS Signal Word: Warning