missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zeolite, type: ZSM-5, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 279571000

 View more versions of this product

Catalog No. AC279571000



Description

Description

Adsorbent
Specifications

Specifications

Zeolite
C14H16N2
1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, 4-2-ethyl-2-indanyl imidazole, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, antisedan, atipamezol spanish, atipamezole, atipamezolum latin, dsstox_cid_29061, unii-03n9u5jaf6
HSWPZIDYAHLZDD-UHFFFAOYSA-N
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole
71310
400 to 570 Molar ratio SiO2/Al2O3
Plastic bottle
Solubility in water: insoluble.
Yellow
308081-08-5
MFCD00132601
Solubility in water: insoluble
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3
212.296
Crystalline Powder
10 % max. (1000°C)
700ppm max.
300 m2/g (BET) min.
100g

Chemical Identifiers

308081-08-5
212.296
HSWPZIDYAHLZDD-UHFFFAOYSA-N
71310
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3
C14H16N2
MFCD00132601
1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, 4-2-ethyl-2-indanyl imidazole, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole, antisedan, atipamezol spanish, atipamezole, atipamezolum latin, dsstox_cid_29061, unii-03n9u5jaf6
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves

GHS Signal Word: Warning

SDS Documents