Alfa Aesar™ Zeolite ferrierite, ammonium
Manufacturer:  Alfa Aesar™ 4588422
 View more versions of this product
Catalog No. AA4588422
Description
Description
  • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene
Specifications
Specifications
Zeolite ferrierite, ammonium
Al2O5Si
SiO2:Al2O3, 20:1 mole ratio
O=[Al]O[Al]=O.O=[Si]=O
162.043
Powder
White
1318-02-1
MFCD00132601
HNPSIPDUKPIQMN-UHFFFAOYSA-N
dioxosilane;oxo(oxoalumanyloxy)alumane
9942228
400m2/g
100g
Chemical Identifiers
1318-02-1
162.043
HNPSIPDUKPIQMN-UHFFFAOYSA-N
dioxosilane;oxo(oxoalumanyloxy)alumane
Al2O5Si
MFCD00132601
9942228
O=[Al]O[Al]=O.O=[Si]=O
SDS Documents