Please login to your online account to display your discounted pricing

Zeolite, Alfa Aesar™

$471.62 - $471.62

Chemical Identifiers

Chemical Name or Material Zeolite
Name Note Hardness: 3.5-5.5; Sp. Gr.: 2.0-2.4. 1 piece 100g. Aluminum silicate with Na, Ca, K and variable amounts of water
CAS 68989-22-0
Physical Form Pieces
MDL Number MFCD00132601
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43628KJ

  SDS
alfa aesar
43628KJ
5 pieces Each for $471.62
Description & Specifications

Specifications

Quantity 5 pieces
Color Pink
Chemical Name or Material Zeolite
Odor Odorless
Name Note Hardness: 3.5-5.5; Sp. Gr.: 2.0-2.4. 1 piece 100g. Aluminum silicate with Na, Ca, K and variable amounts of water
Physical Form Pieces
MDL Number MFCD00132601
CAS 68989-22-0