Please login to your online account to display your discounted pricing

Alfa Aesar™ Zeocin™, 100mg/mL in HEPES, sterile-filtered

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material Zeocin™
Name Note 100mg/mL in HEPES, sterile-filtered
MDL Number MFCD28100499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ671408EQ

  SDS
alfa aesar™
J671408EQ
5 x 1mL Pack for $213.96
Description & Specifications

Specifications

Chemical Name or Material Zeocin™
MDL Number MFCD28100499
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Storage Note 1 -30° to -10°C
Name Note 100mg/mL in HEPES, sterile-filtered
Quantity 5 x 1mL
Health Hazard 1 Warning

Applications
Zeocin, a popular and effective antibiotic for the selection of vectors bearing the Sh ble gene in a variety of cells types (bacteria, eukaryotic microorganisms, plant and animal cells). Resistance to Zeocin is conferred by the Sh ble gene product, which inactivates Zeocin™ by binding to the antibiotic. Sh ble is a small gene, only 370bp in size, carried in a number of InvivoGen’s vectors. Zeocin is used as a selective agent in molecular genetics experiments.

Solubility
Fully miscible.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Supplied in 10mM HEPES, pH 7.5. High ionic strength or basicity inhibits the activity of zeocin. Use at pH of approx. 7.5. For selection of E. coli concentration of NaCl should not exceed 5g/L. Zeocin is a trademark of Cayla

Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.

-30° to -10°C