missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zein, purified, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 179315000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zein | |
9010-66-6 | |
Plastic bottle | |
500 g | |
Solubility in water: insoluble. Other solubilities: soluble in aq. solutions ph >12,aq. ethanol,,soluble in ethanol,dmf |
Purified | |
2% max. (800°C) | |
MFCD00062414 | |
14, 10119 |
Chemical Identifiers
9010-66-6 | |
MFCD00062414 |
Safety and Handling
EINECSNumber : 232-722-9
RUO – Research Use Only