missing translation for 'onlineSavingsMsg'
Learn More

Zearalenone Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018137
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018137 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018137 Supplier Merck Emd Millipore Supplier No. 341262ML
Only null left

Zearalenone is a mycotoxin produced byFusarium graminearum, culmorum crookwellense and roseum in food of plant origin. It is an endocrine disrupter since it binds to the estrogen receptors and interacts with the steroidogenic enzymes.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 17924-92-4
Linear Formula C18H22O5
MDL Number MFCD00133085
Synonym ZON
Type Analytical Standard
Quantity 2 mL
Formula Weight 318.36
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile