Please login to your online account to display your discounted pricing

ZD 7288, Tocris


Sino-atrial node function modulator (If inhibitor)

Quantity 10mg

Manufacturer: r & d systems  100010

 View more versions of this product

Catalog No. 100010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 292.81
Quantity 10mg
Molecular Formula C15H21ClN4
Synonym 4-Ethylphenylamino-1, 2-dimethyl-6-methylaminopyrimidinium chloride
Recommended Storage Desiccate at +4°C
Chemical Name or Material ZD 7288
Assay Percent Range >99%
CAS 133059-99-1

Specifications

Chemical Name or Material ZD 7288
Assay Percent Range >99%
Molecular Formula C15H21ClN4
Formula Weight 292.81
Synonym 4-Ethylphenylamino-1, 2-dimethyl-6-methylaminopyrimidinium chloride
Accessories

Desiccate at +4°C