Please login to your online account to display your discounted pricing

ZD 7288, Tocris

Sino-atrial node function modulator (If inhibitor)

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material ZD 7288
Assay Percent Range >99%
Molecular Formula C15H21ClN4
Formula Weight 292.81
Synonym 4-Ethylphenylamino-1, 2-dimethyl-6-methylaminopyrimidinium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

100010

 
r & d systems
100010
10mg Each for $195.49

100050

 
r & d systems
100050
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 292.81
Quantity 10mg
Molecular Formula C15H21ClN4
Synonym 4-Ethylphenylamino-1, 2-dimethyl-6-methylaminopyrimidinium chloride
Recommended Storage Desiccate at +4°C
Chemical Name or Material ZD 7288
Assay Percent Range >99%
CAS 133059-99-1
Safety and Handling

Desiccate at +4°C