Please login to your online account to display your discounted pricing

ZD 7114 hydrochloride, Tocris

β3 agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material ZD 7114 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H30N2O6·HCl
Formula Weight 454.95
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]-N-(2-methoxyethyl)phenoxyacetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

093010

 
r & d systems
RDS093010
10mg Each for $205.70

093050

 
r & d systems
RDS093050
50mg Each for $862.95
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 454.95
Chemical Name or Material ZD 7114 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H30N2O6·HCl
CAS 129689-28-7
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]-N-(2-methoxyethyl)phenoxyacetamide hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature