Please login to your online account to display your discounted pricing

ZCL 278, Tocris™

Cdc42 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material ZCL 278
Assay Percent Range >95%
Molecular Formula C21H19BrClN5O4S2
Formula Weight 584.89
Synonym 2-(4-Bromo-2-chlorophenoxy)-N-[[[4-[[(4, 6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]thioxomethyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

479410

 
r & d systems
RDS479410
10mg Each for $252.39

479450

 
r & d systems
RDS479450
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 584.89
Quantity 10mg
Recommended Storage Store at +4°C
CAS 587841-73-4
Chemical Name or Material ZCL 278
Assay Percent Range >95%
Synonym 2-(4-Bromo-2-chlorophenoxy)-N-[[[4-[[(4, 6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]thioxomethyl]acetamide
Molecular Formula C21H19BrClN5O4S2
Safety and Handling

Store at +4°C