Please login to your online account to display your discounted pricing

Zardaverine, Tocris


PDE3/4 inhibitor

Quantity 10mg

Manufacturer: r & d systems  104610

 View more versions of this product

Catalog No. 104610

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 268.22
Quantity 10mg
Chemical Name or Material Zardaverine
Assay Percent Range >98%
Molecular Formula C12H10F2N2O3
CAS 101975-10-4
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Zardaverine
Assay Percent Range >98%
Molecular Formula C12H10F2N2O3
Formula Weight 268.22
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
Accessories

Store at Room Temperature