Please login to your online account to display your discounted pricing

Zardaverine, Tocris

PDE3/4 inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Zardaverine
Assay Percent Range >98%
Molecular Formula C12H10F2N2O3
Formula Weight 268.22
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

104650

 
r & d systems
104650
50mg Each for $682.67

104610

 
r & d systems
104610
10mg Each for $164.61
Description & Specifications

Specifications

Formula Weight 268.22
Quantity 50mg
Molecular Formula C12H10F2N2O3
Assay Percent Range >98%
Chemical Name or Material Zardaverine
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
Recommended Storage Store at Room Temperature
CAS 101975-10-4
Safety and Handling

Store at Room Temperature