Please login to your online account to display your discounted pricing

Zardaverine, Tocris


PDE3/4 inhibitor

Quantity 50mg

Manufacturer: r & d systems  104650

 View more versions of this product

Catalog No. 104650

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Zardaverine
CAS 101975-10-4
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C12H10F2N2O3
Formula Weight 268.22
Quantity 50mg
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone

Specifications

Chemical Name or Material Zardaverine
Assay Percent Range >98%
Molecular Formula C12H10F2N2O3
Formula Weight 268.22
Synonym 6-[4-(Difluoromethoxy)-3-methoxyphenyl]-3(2H)-pyridazinone
Accessories

Store at Room Temperature