Please login to your online account to display your discounted pricing

ZAPA sulfate, Tocris

Agonist at “low affinity” GABAA receptor; More potent than GABA/muscimol

$159.50 - $678.85

Chemical Identifiers

Chemical Name or Material ZAPA sulfate
Assay Percent Range >98%
Molecular Formula C4H6N2O2S·H2SO4
Formula Weight 244.24
Synonym (Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

018050

 
r & d systems
RDS018050
50mg Each for $678.85

018010

 
r & d systems
RDS018010
10mg Each for $159.50
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 244.24
Chemical Name or Material ZAPA sulfate
CAS 371962-01-5
Assay Percent Range >98%
Molecular Formula C4H6N2O2S·H2SO4
Synonym (Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature