missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human ZAP-70, His Tag Recombinant Protein
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins.
$1308.00 - $2645.00
Specifications
Accession Number | NP_001070 |
---|---|
Gene Alias | ZAP-70 |
KinaseFamily | SYK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Description
ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation.
Specifications
NP_001070 | |
SYK Kinase Family | |
Recombinant | |
Baculovirus |
ZAP-70 | |
Human | |
His Tag |