Please login to your online account to display your discounted pricing

Zacopride hydrochloride, Tocris

Highly potent 5-HT3 receptor antagonist; Also 5-HT4 agonist

$179.63 - $750.85

Chemical Identifiers

Chemical Name or Material Zacopride hydrochloride
Assay Percent Range >99%
Molecular Formula C15H20ClN3O2·HCl
Formula Weight 346.26
Synonym (±)-4-Amino-N-1-azabicyclo[2.2.2]oct-3-yl-5-chloro-2-methoxybenzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

179510

 
r & d systems
RDS179510
10mg Each for $179.63

179550

 
r & d systems
RDS179550
50mg Each for $750.85
Description & Specifications

Specifications

Formula Weight 346.26
Quantity 10mg
Chemical Name or Material Zacopride hydrochloride
Assay Percent Range >99%
Molecular Formula C15H20ClN3O2·HCl
CAS 101303-98-4
Synonym (±)-4-Amino-N-1-azabicyclo[2.2.2]oct-3-yl-5-chloro-2-methoxybenzamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature