missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (Z)-Stilbenediboronic acid bis(pinacol) ester, 98%

$99.04 - $321.62

Chemical Identifiers

CAS 221006-76-4
Molecular Formula C26H34B2O4
Formula Weight 432.18
MDL Number MFCD02683500
Beilstein 9081275
Synonym cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)stilbene
Chemical Name or Material (Z)-Stilbenediboronic acid bis(pinacol) ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1965203 SDS Alfa Aesar™
L1965203
1g Each for $99.04
AAL1965206 SDS Alfa Aesar™
L1965206
5g Each for $321.62
Specifications Description & Specifications

Chemical Identifiers

221006-76-4
432.18
9081275
(Z)-Stilbenediboronic acid bis(pinacol) ester
C26H34B2O4
MFCD02683500
cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)stilbene

Specifications

221006-76-4
176° to 179°C
MFCD02683500
9081275
(Z)-Stilbenediboronic acid bis(pinacol) ester
C26H34B2O4
432.18
1g
cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)stilbene
98%