missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Z-Guggulsterone

Broad spectrum steroid receptor ligand. More active isomer of guggulsterone (Cat. No. 2013)

$250.58 - $1,042.86

Specifications

Product Type Z-Guggulsterone
Inhibitors Z-Guggulsterone
Molecular Weight 312.45
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
357010R Tocris™
RDS357010
10mg Each for $250.58
357050R Tocris™
RDS357050
50mg Each for $1,042.86
Specifications

Description

Broad spectrum steroid receptor ligand; mineralocorticoid, progesterone and glucocorticoid receptor antagonist (Ki values are 37, 224 and 252nM respectively) and weak androgen receptor agonist (Ki = 315nM). Induces apoptosis in prostate cancer cells and inhibits angiogenesis via suppression of the VEGF-VEGFR2-Akt signaling pathway. Exhibits antilipidemic activity via antagonism of the farnesoid X receptor (FXR) and displays antiseptic, antirheumatic and anti-inflammatory activity in vivo. More active isomer of guggulsterone (Cat. No. 2013).
  • Chemical Name: (Z)-Pregna-4,17(20)-diene-3,16-dione
  • Formula: C21H28O2
  • CAS Number: 39025-23-5
  • Solubility: Soluble to 25mM in DMSO and to 10mM in ethanol with gentle warming
  • Purity: >98
Description & Specifications

Specifications

Z-Guggulsterone
Z-Guggulsterone
312.45
>98