Please login to your online account to display your discounted pricing

Novus Biologicals™ Z-FA-FMK Inhibitor


For use in research applications

Manufacturer: novus biologicals™  NBP2293845MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2293845

  • / Each
 

Description & Specifications

Specifications

Inhibitors Z-FA-FMK
Quantity 5mg
Product Type Z-FA-FMK Inhibitor
Molecular Weight 386kd
For Use With (Application) Inhibits of cathepsins B and L
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.