missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Z-FA-FMK Inhibitor

For use in research applications

$259.02 - $442.22

Specifications

Product Type Z-FA-FMK Inhibitor
Inhibitors Z-FA-FMK
Molecular Weight 386kd
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
For Use With (Application) Inhibits of cathepsins B and L
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP2293843 Novus Biologicals™
NBP2293843MG
3mg Each for $259.02
NBP2293845 Novus Biologicals™
NBP2293845MG
5mg Each for $442.22
Specifications Description & Specifications

Specifications

Z-FA-FMK Inhibitor
386kd
Inhibits of cathepsins B and L
Z-FA-FMK
Store at -20°C. Avoid freeze-thaw cycles.