Please login to your online account to display your discounted pricing

Z-Cyclopentyl-AP4, Tocris

Group III mGlu agonist (mGlu4 > mGlu8 > mGlu7)

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Z-Cyclopentyl-AP4
Assay Percent Range >98%
Molecular Formula C6H12NO5P
Formula Weight 209.14
Synonym cis-(±)-1-Amino-3-phosphonocyclopentane carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

324910

 
r & d systems
RDS324910
10mg Each for $239.63

324950

 
r & d systems
RDS324950
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 209.14
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C6H12NO5P
Chemical Name or Material Z-Cyclopentyl-AP4
Synonym cis-(±)-1-Amino-3-phosphonocyclopentane carboxylic acid
CAS 103439-17-4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C