Please login to your online account to display your discounted pricing

(Z)-4-Hydroxytamoxifen, Tocris™

Metabolite of tamoxifen

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material (Z)-4-Hydroxytamoxifen
Assay Percent Range >98%
Molecular Formula C26H29NO2
Formula Weight 387.51
Synonym 4-[(1Z)-1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

341250

 
r & d systems
RDS341250
50mg Each for $924.23

34-1210

 
r & d systems
RDS341210
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 387.51
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C26H29NO2
CAS 68047-06-3
Synonym 4-[(1Z)-1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol
Recommended Storage Store at +4°C
Chemical Name or Material (Z)-4-Hydroxytamoxifen
Safety and Handling

Store at +4°C