Please login to your online account to display your discounted pricing

Tocris™ YZ9

PFKFB3 inhibitor

$113.57 - $478.51

Specifications

Product Type YZ9
Inhibitors YZ9
Molecular Weight 234.2
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

504810R

 
tocris™
RDS504810
10mg Each for $113.57

504850R

 
tocris™
RDS504850
50mg Each for $478.51
Description & Specifications

Specifications

Product Type YZ9
Inhibitors YZ9
Molecular Weight 234.2
Purity >98

PFKFB3 (6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase) inhibitor (IC50 = 0.183μM). Also competitively inhibits F-6-P (Ki = 0.094μM). Inhibits cell growth (GI50 = 2.7μM) in a XTT-based in vitro toxicology assay.

  • Chemical Name: Ethyl 7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylate
  • Formula: C12H10O5
  • CAS Number: 6093-71-6
  • Solubility: Soluble to 50mM in DMSO and to 20mM in ethanol
  • Purity: >98