Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium tris(trifluoromethylsulfonyl)imide

Chemical Identifiers

Chemical Name or Material Yttrium tris(trifluoromethylsulfonyl)imide
CAS 189114-86-1
Linear Formula [(CF3SO2)2N]3Y
Formula Weight 929.31
MDL Number MFCD23380178
UN Number UN3261
Synonym Yttrium tris(trifluoromethanesulfonyl)imide; Yttrium tris(trifluoromethanesulfonimide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4719303

  SDS
alfa aesar™
4719303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4719306

  SDS
alfa aesar™
4719306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47193MD

  SDS
alfa aesar™
47193MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 929.31
Chemical Name or Material Yttrium tris(trifluoromethylsulfonyl)imide
Physical Form Solid
Sensitivity Hygroscopic
Linear Formula [(CF3SO2)2N]3Y
MDL Number MFCD23380178
CAS 189114-86-1
UN Number UN3261
Synonym Yttrium tris(trifluoromethanesulfonyl)imide; Yttrium tris(trifluoromethanesulfonimide)
Color Pale yellow
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.