missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040300DJ

 View more versions of this product

Catalog No. AA40300DJ


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Yttrium rod
7440-65-5
Y
50 mm
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
99.9%
Mylar under Argon
Cast Rod
12.7mm (0.5 in.) dia.
(metals basis excluding Ta)
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Odorless
≈0.57g/mm

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]
Safety and Handling

Safety and Handling

EINECSNumber : 231-174-8

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only