missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium Powder, -40 mesh, 0.999, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4414714

 View more versions of this product

Catalog No. AA4414714



Specifications

Specifications

Yttrium
Rare Earth Oxides Content
Gray powder
99.90%
MFCD00011468
element
[Y]
Odorless
Under Argon
CHEBI:33331
Gray
25g
7440-65-5
4.47g/cm3
Y
-40 Mesh
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
88.906
23993
Powder

Chemical Identifiers

7440-65-5
88.906
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
element
CHEBI:33331
[Y]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-174-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents