missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium, plasma standard solution, Y 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13855AD

 View more versions of this product

Catalog No. AA13855AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Yttrium
Liquid
MFCD00011468
Liquid
50mL
1314-36-9,1314-36-9,7732-18-5
Y2O3 in 5% HNO3
Nitric acid: 5.0%; Yttrium(III) oxide: 0.127%; Water: 94.873%
23 hPa (17mm Hg) at 20°C

Chemical Identifiers

1314-36-9
Y2O3 in 5% HNO3
MFCD00011468
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents