missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium, plasma standard solution, Specpure™ Y 1000μg/mL, Thermo Scientific Chemicals

$123.00 - $301.73

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13855AE
View Documents Promotion Details
Thermo Scientific Chemicals
013855AE
100 mL
Each for $123.00
 
AA13855AP
View Documents Promotion Details
Thermo Scientific Chemicals
013855AP
500 mL
Each for $301.73
 
Description

Description

Yttrium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide

Specifications

1314-36-9,7732-18-5
O3Y2
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
Nitric acid: 5.0%; Yttrium(III) oxide: 0.127%; Water: 94.873%
Miscible with water
Yttrium
100 mL
MFCD00011473
[O--].[O--].[O--].[Y+3].[Y+3]
225.81
23 hPa (17mm Hg) at 20°C
Liquid
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8, Packing Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only