Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium, Oil based standard solution, Specpure™, Y 1000μg/g

Chemical Identifiers

Chemical Name or Material Yttrium, Oil based standard solution
Name Note Y 1000μg/mL
Purity Grade Specpure™
MDL Number MFCD00011468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4717218

  SDS
alfa aesar™
4717218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Chemical Name or Material Yttrium, Oil based standard solution
Purity Grade Specpure™
Name Note Y 1000μg/mL
MDL Number MFCD00011468
TSCA TSCA
Physical Form Liquid