missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) trifluoroacetate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4459709

 View more versions of this product

Catalog No. AA4459709



Specifications

Specifications

Yttrium (III) trifluoroacetate hydrate
(REO)
MFCD05663780
Insoluble in water.
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.O.[Y]
448.99
427.95 (Anhydrous)
99.9%
304851-95-4
C6H5F9O7Y
yttrium trifluoroacetate
RTYFWTAMUWPTNO-UHFFFAOYSA-N
2,2,2-trifluoroacetic acid;yttrium;hydrate
22836421
Crystalline
10g

Chemical Identifiers

304851-95-4
448.99
RTYFWTAMUWPTNO-UHFFFAOYSA-N
22836421
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.O.[Y]
C6H5F9O7Y
MFCD05663780
yttrium trifluoroacetate
2,2,2-trifluoroacetic acid;yttrium;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents