Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Chemical Identifiers

Chemical Name or Material Yttrium(III) perchlorate
CAS 14017-56-2
Purity Grade Reagent Grade
Formula Weight 495.34 (387.25 Anhydrous)
MDL Number MFCD00150430
UN Number UN3211
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40580NB

 
alfa aesar™
40580NB
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40580ND

 
alfa aesar™
40580ND
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 495.34 (387.25 Anhydrous)
Chemical Name or Material Yttrium(III) perchlorate
Purity Grade Reagent Grade
Physical Form Liquid
MDL Number MFCD00150430
CAS 14017-56-2
UN Number UN3211
Color Colorless
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H314
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 50% w/w aq. soln.
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H314
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.