Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Magnetron sputtering target

Chemical Identifiers

Chemical Name or Material Yttrium(III) oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 225.81
MDL Number MFCD00011473
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41119KS

  SDS
alfa aesar™
41119KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.81
Chemical Name or Material Yttrium(III) oxide sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011473
Quantity 1 each