missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) oxide, REacton™, nanopowder, 99.995% (REO)

Manufacturer:  Alfa Aesar™ 4404822

 View more versions of this product

Catalog No. AA4404822



Specifications

Specifications

Yttrium (III) oxide
(REO)
MFCD00011473
Insoluble in water. Soluble in alcohol and dilute acids.
25-50nm APS Powder
Reacton™
Odorless
Air Sensitive
2410°C
1314-36-9
Y2O3
14,10107
225.81
99.995%
5.01 g/mL
25 to 50nm APS
White
100g

Chemical Identifiers

1314-36-9
Y2O3
MFCD00011473
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-233-5

RTECSNumber : ZG3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents