Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) oxide, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Yttrium(III) oxide
CAS 1314-36-9
Purity Grade Reacton™
Formula Weight 225.81
MDL Number MFCD00011473
Merck Index 14,10107
Synonym Yttria
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA66178A1

 
alfa aesar™
66178A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Reacton™
Formula Weight 225.81
Chemical Name or Material Yttrium(III) oxide
TSCA TSCA
Odor Odorless
Physical Form Powder
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011473
CAS 1314-36-9
Merck Index 14,10107
Synonym Yttria
Melting Point 2410°C
Quantity 1kg
Density 5.01g/mL
RTECS Number ZG3850000