Learn More
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals
Sintered pellets suitable for vacuum deposition
Supplier: Thermo Scientific Chemicals 4064836
Description
Sintered pellets suitable for vacuum depositionYttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is a precursor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yttrium(III) oxide pellets | |
approx. 10mm dia. x 5mm thick | |
500 g | |
O3Y2 | |
14,10107 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
diyttrium(3+) trioxidandiide | |
225.81 | |
Odorless | |
5.01 g/mL |
2410°C | |
1314-36-9 | |
(REO) | |
MFCD00011473 | |
Insoluble in water. Soluble in alcohol and dilute acids. | |
[O--].[O--].[O--].[Y+3].[Y+3] | |
225.81 | |
99.9% | |
Air Sensitive |
Chemical Identifiers
1314-36-9 | |
225.81 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Y+3].[Y+3] |
O3Y2 | |
MFCD00011473 | |
diyttrium(3+) trioxidandiide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-233-5
RTECSNumber : ZG3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only