Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) oxide pellets, 99.9% (REO)

Sintered pellets suitable for vacuum deposition

Chemical Identifiers

Chemical Name or Material Yttrium(III) oxide pellets
CAS 1314-36-9
Name Note approx. 10mm dia. x 5mm thick
Formula Weight 225.81
MDL Number MFCD00011473
Merck Index 14,10107
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4064822

  SDS
alfa aesar™
4064822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4064836

  SDS
alfa aesar™
4064836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.81
Odor Odorless
Melting Point 2410°C
Quantity 100g
Assay Percent Notes (REO)
Assay 99.9%
CAS 1314-36-9
Density 5.01g/mL
Merck Index 14,10107
TSCA TSCA
Sensitivity Air Sensitive
RTECS Number ZG3850000
Name Note approx. 10mm dia. x 5mm thick
Chemical Name or Material Yttrium(III) oxide pellets
MDL Number MFCD00011473