missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Yttrium(III) oxide, nanopowder, 99.995% (REO)

Manufacturer:  Alfa Aesar™ 4715414

 View more versions of this product

Catalog No. AA4715414



Specifications

Specifications

Yttrium(III) oxide
(REO)
MFCD00011473
225.81
99.995%
Odorless
White
25g
1314-36-9
Y2O3
14,10107
Nanopowder
5.01g/mL
50 to 70nm APS
2410°C

Chemical Identifiers

1314-36-9
MFCD00011473
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : ZG3850000

TSCA : TSCA

SDS Documents