Learn More
Yttrium(III) oxide, nanopowder, 99.995% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04715436
Description
Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yttrium(III) oxide | |
1314-36-9 | |
Powder | |
(REO) | |
Y2O3 | |
14,10107 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
diyttrium(3+) trioxidandiide | |
225.81 | |
Odorless |
2410°C | |
White | |
500 g | |
O3Y2 | |
MFCD00011473 | |
Insoluble in water. Soluble in alcohol and dilute acids | |
[O--].[O--].[O--].[Y+3].[Y+3] | |
225.81 | |
99.995% | |
5.01 g/mL |
Chemical Identifiers
1314-36-9 | |
225.81 | |
RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Y+3].[Y+3] |
O3Y2 | |
MFCD00011473 | |
diyttrium(3+) trioxidandiide |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-233-5
RTECSNumber : ZG3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only