Please login to your online account to display your discounted pricing

Yttrium(III) oxide, 99.9999% (REO), Alfa Aesar™ Reacton™

$197.39 - $2,456.01

Chemical Identifiers

Chemical Name or Material Yttrium(III) oxide
CAS 1314-36-9
Physical Form Powder
Assay Percent Range 99.9999% (REO)
Molecular Formula Y2O3
Formula Weight 225.81
MDL Number MFCD00011473
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4286409

  SDS
alfa aesar
4286409
10g Each for $197.39

AA4286418

  SDS
alfa aesar
4286418
50g Each for $658.46

AA4286430

  SDS
alfa aesar
4286430
250g Each for $2,456.01
Description & Specifications

Specifications

Formula Weight 225.81
Quantity 10g
Physical Form Powder
Density 5.01 g/mL
EINECS Number 215-233-5
CAS 1314-36-9
Chemical Name or Material Yttrium(III) oxide
Assay Percent Range 99.9999% (REO)
Molecular Formula Y2O3
MDL Number MFCD00011473