missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium(III) oxide, 99.99%, (trace metal basis), ACROS Organics™

Manufacturer:  Fisher Scientific Acros 194610500

 View more versions of this product

Catalog No. AC194610500



Description

Description

for ultra trace analysis
Specifications

Specifications

Yttrium(III)oxide
99.99% min. (Y2O3) TREO base
Y2O3
14, 10107
225.81
Crystalline Powder
Trace Metal Basis
Glass bottle
Solubility in water: insoluble. Other solubilities: soluble in dilute acids
White
50g
1314-36-9
O3Y2
MFCD00011473
Solubility in water: insoluble. Other solubilities: soluble in dilute acids
225.81
99.99%
1.0 % max. (1000°C) (material base)
2 to 10 Micron
100ppm max.
2410.0°C

Chemical Identifiers

1314-36-9
O3Y2
225.81
MFCD00011473
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-233-5

SDS Documents