missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1118122

 View more versions of this product

Catalog No. AA1118122



Specifications

Specifications

Yttrium(III) oxide
99.99% (REO)
MFCD00011473
≪10 Micron Powder
100g
1314-36-9
O3Y2
225.81
5.01g/mL

Chemical Identifiers

1314-36-9
O3Y2
MFCD00011473
Safety and Handling

Safety and Handling

EINECSNumber : 215-233-5

Documents